This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03945207_s1
See other ATP6V1B2 GE Assays ›
Gene Symbol:
Atp6v1b2
Gene Name:
ATPase, H+ transporting, lysosomal V1 subunit B2
Gene Aliases:
AI194269, AI790362, Atp6b2, HO57, R74844
Chromosome Location:
Chr.8: 69088736 - 69113718 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03945207_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Atp6v1b2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK079830.1 - 1 1252 84
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BE628878.1 - 2 188 Chr. 8 - 69088789 - 69096518 [+] on Build GRCm38 84
GenBankmRNA BG094715.1 - 2 131 Chr. 8 - 69088849 - 69096586 [+] on Build GRCm38 84

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream solute carrier family 18 (vesicular monoamine),
member 1
Slc18a1 4832416I10Rik,
Vat1,
Vmat1
-4.4kb Anti-sense Chr. 8 - 69037708 - 69090629 on NCBI Build GRCm38
Downstream leucine zipper,
putative tumor suppressor 1
Lzts1 F37,
FEZ1
35.8kb Anti-sense Chr. 8 - 69132669 - 69184220 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0