This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03944732_s1
See other LOC497255 GE Assays ›
Gene Symbol:
LOC497255
Gene Name:
uncharacterized LOC497255
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03944732_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC497255
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK078348.1 - 1 646 81
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AI415469.1 - 1 Chr. 15 - 73423653 - 73424111 [-] on Build GRCm38 81

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream PTK2 protein tyrosine kinase 2 Ptk2 FAK,
FRNK,
Fadk,
mKIAA4203,
p125FAK
-36.0 Anti-sense Chr. 15 - 73205102 - 73423275 on NCBI Build GRCm38
Downstream DENN/MADD domain containing 3 Dennd3 AI447457,
E030003N15Rik
88.5kb Sense Chr. 15 - 73512560 - 73572242 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix ABI

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2288645 AK078348.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 102071 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30330364 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30330347 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30330393 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30330390 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30330385 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30330389 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12324284 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12436179 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30330375 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30330365 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12297511 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30330382 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12436180 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30330345 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P11966550 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30330344 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30330384 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30330392 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30330357 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P11887250 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12174864 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30330355 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30330348 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30330398 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30330367 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30330401 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30330406 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12174863 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30330352 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30330396 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30330388 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P11966551 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30330353 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12297514 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30330359 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30330395 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30330403 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12297505 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30330350 - -
Array Name Probe IDs Targets Target Type
Affymetrix HT MG-430 PM Array Plate 1459678_PM_at - -
Mouse Genome 430 2.0 Array 1459678_PM_at - -
Mouse Genome 430 2.0 Array 1459678_at - -
Mouse Expression Set 430 1459678_at - -
Mouse Expression Set 430 1459678_PM_at - -
Murine Genome U74v2 Set 131556_at - -
Murine Genome U74v2 Set 131556_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1459678_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1459678_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 708091 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0