This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03944483_s1
See other ACTA2 GE Assays ›
Gene Symbol:
Acta2
Gene Name:
actin, alpha 2, smooth muscle, aorta
Gene Aliases:
0610041G09Rik, Actvs, SMalphaA, a-SMA, alphaSMA
Chromosome Location:
Chr.19: 34240336 - 34255585 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03944483_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Acta2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_007392.3 NP_031418.1 9 1970 102
GenBank mRNA AK077294.1 - 1 49 102
AK080187.1 - 1 603 102
AK137170.1 - 1 610 102
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BB222563.2 - 1 118 Chr. 19 - 34240388 - 34241053 [-] on Build GRCm38 102
GenBankmRNA BY716908.1 - 47 Chr. - - [-] on Build 102

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream ankyrin repeat domain 22 Ankrd22 5430429D21Rik,
D19Ertd675e
-74.3kb Sense Chr. 19 - 34122549 - 34166041 on NCBI Build GRCm38
Downstream Fas (TNF receptor superfamily member 6) Fas AI196731,
APO1,
APT1,
CD95,
TNFR6,
Tnfrsf6,
lpr
49.7kb Anti-sense Chr. 19 - 34290659 - 34327775 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Acta2

Panther Classification:

Molecular Function -

actin and actin related protein actin or actin-binding cytoskeletal protein cytoskeletal protein

Gene Ontology Categories:

Function(s) Process(es)

muscle contraction
regulation of blood pressure
vascular smooth muscle contraction
nucleotide binding
protein binding
ATP binding
protein kinase binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1900012395.mm.1 NM_007392.3 Refseq
Affymetrix Mouse Clariom S Array PSR1900012395.mm.1 TC1900001415.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 325295 - -
ABI Mouse Genome Survey Microarray 724403 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0