This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03944382_s1
See other 4930455C13RIK GE Assays ›
Gene Symbol:
4930455C13Rik
Gene Name:
RIKEN cDNA 4930455C13 gene
Gene Aliases:
-
Chromosome Location:
Chr.10: 21319088 - 21342055 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03944382_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 4930455C13Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK076940.1 - 1 549 101
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BX639191.1 - 1 Chr. 10 - 21341364 - 21341595 [+] on Build GRCm38 101
GenBankmRNA BB017040.2 - 1 558 Chr. 10 - 21341365 - 21342028 [-] on Build GRCm38 101

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream predicted gene,
33619
Gm33619 -152.7kb Anti-sense Chr. 10 - 21186921 - 21188618 on NCBI Build GRCm38
Downstream aldehyde dehydrogenase 8 family,
member A1
Aldh8a1 Raldh4 35.3kb Anti-sense Chr. 10 - 21377300 - 21396578 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header