This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03943327_s1
See other D130067P18RIK GE Assays ›
Gene Symbol:
D130067P18Rik
Gene Name:
RIKEN cDNA D130067P18 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03943327_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: D130067P18Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK051729.1 - 1 156 99
AK051977.1 - 1 155 99
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BB658080.1 - 1 155 Chr. 12 - 108579139 - 108579765 [+] on Build GRCm38 99
GenBankmRNA BB656927.1 - 1 156 Chr. 12 - 108579139 - 108579787 [+] on Build GRCm38 99

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream predicted gene,
16596
Gm16596 -23.5kb Anti-sense Chr. 12 - 108536402 - 108555618 on NCBI Build GRCm38
Downstream microRNA 342 Mir342 Mirn342,
mmu-mir-342
77.1kb Sense Chr. 12 - 108658620 - 108658718 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

MicroArray Probes:

Agilent

Array Name Probe IDs Targets Target Type
Whole Mouse Genome G4122A A_52_P588399 AK051729.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P588399 AK051977.1 mouse_transcriptome

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0