This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03942516_s1
See other 8430401P03RIK GE Assays ›
Gene Symbol:
8430401P03Rik
Gene Name:
RIKEN cDNA 8430401P03 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03942516_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 8430401P03Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK018374.1 - 1 849 77

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream inhibin beta-C Inhbc -23.6kb Anti-sense Chr. 10 - 127356325 - 127370544 on NCBI Build GRCm38
Downstream SH3 and cysteine rich domain 3 Stac3 9830125E18 106.4kb Sense Chr. 10 - 127501617 - 127508823 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Six_ChIP_02_of_03_v2 A_68_P08377616 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27535604 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27535581 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27535582 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27535596 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27535597 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27535620 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27535607 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27535600 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27535583 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27535606 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27535590 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27535585 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27535592 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27535611 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27535599 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08861868 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27535588 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27535594 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27535584 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08549836 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27535601 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08402726 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08332319 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27535609 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27535591 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08669513 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27535621 - -
Array Name Probe IDs Targets Target Type
Affymetrix HT MG-430 PM Array Plate 1433367_PM_at - -
Affymetrix HT MG-430 PM Array Plate 1433369_PM_at - -
Affymetrix HT MG-430 PM Array Plate 1433368_PM_x_at - -
Mouse Genome 430 2.0 Array 1433367_at - -
Mouse Genome 430 2.0 Array 1433369_PM_at - -
Mouse Genome 430 2.0 Array 1433368_x_at - -
Mouse Genome 430 2.0 Array 1433369_at - -
Mouse Genome 430 2.0 Array 1433368_PM_x_at - -
Mouse Genome 430 2.0 Array 1433367_PM_at - -
Mouse Expression Set 430 1433369_PM_at - -
Mouse Expression Set 430 1433369_at - -
Mouse Expression Set 430 1433367_PM_at - -
Mouse Expression Set 430 1433367_at - -
Mouse Expression Set 430 1433368_x_at - -
Mouse Expression Set 430 1433368_PM_x_at - -
Affymetrix HT Mouse Genome 430B Array 1433369_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1433368_PM_x_at - -
Affymetrix HT Mouse Genome 430B Array 1433367_at - -
Affymetrix HT Mouse Genome 430B Array 1433369_at - -
Affymetrix HT Mouse Genome 430B Array 1433367_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1433368_x_at - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 102260427 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header