This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03942154_s1
See other 6330417A16RIK GE Assays ›
Gene Symbol:
6330417A16Rik
Gene Name:
RIKEN cDNA 6330417A16 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03942154_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 6330417A16Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK018186.1 - 1 1251 99

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream RIKEN cDNA E430025E21 gene E430025E21Rik AL022848,
C76463,
Kiaa0196,
mKIAA0196,
strumpellin
-112.7kb Anti-sense Chr. 15 - 59331997 - 59374194 on NCBI Build GRCm38
Downstream tribbles homolog 1 (Drosophila) Trib1 A530090O15Rik,
TRB-1,
Trb1
160.0kb Sense Chr. 15 - 59648654 - 59657099 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Affymetrix HT MG-430 PM Array Plate 1429996_PM_at - -
Mouse Genome 430 2.0 Array 1429996_PM_at - -
Mouse Genome 430 2.0 Array 1429996_at - -
Mouse Expression Set 430 1429996_at - -
Mouse Expression Set 430 1429996_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1429996_at - -
Affymetrix HT Mouse Genome 430B Array 1429996_PM_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 446158 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 104670601 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0