This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03942075_s1
See other 6330407A03RIK GE Assays ›
Gene Symbol:
6330407A03Rik
Gene Name:
RIKEN cDNA 6330407A03 gene
Gene Aliases:
-
Chromosome Location:
Chr.4: 3714964 - 3716813 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03942075_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 6330407A03Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_028126.1 1 1452 78
GenBank mRNA AK018133.1 - 1 1450 78
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AW519657.1 - 1 Chr. 4 - 3716277 - 3716820 [-] on Build GRCm38 78
GenBankmRNA BX527408.1 - 1 Chr. 4 - 3716317 - 3716829 [-] on Build GRCm38 78

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream Yamaguchi sarcoma viral (v-yes-1) oncogene homolog Lyn AA407514,
Hck-2,
p53Lyn,
p56Lyn
Sense Chr. 4 - 3678121 - 3791612 on NCBI Build GRCm38
Downstream ribosomal protein S20 Rps20 4632426K06Rik,
Dsk4
117.7kb Anti-sense Chr. 4 - 3834473 - 3835600 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2238406 AK018133.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2238406 NR_028126.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P407515 AK018133.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P407515 NR_028126.1 mouse_transcriptome
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10503108 - -
Affymetrix HT MG-430 PM Array Plate 1431374_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10503107 - -
Mouse Genome 430 2.0 Array 1431374_PM_at - -
Mouse Genome 430 2.0 Array 1431374_at - -
Affymetrix Mouse Clariom S Array PSR0400000116.mm.1 NR_028126.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400000116.mm.1 NR_028126.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400000116.mm.1 TC0400000016.mm.1 MTA TC
Mouse Expression Set 430 1431374_at - -
Mouse Expression Set 430 1431374_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1431374_at - -
Affymetrix HT Mouse Genome 430B Array 1431374_PM_at - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 103830114 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0