This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03941774_s1
See other 5830434F19RIK GE Assays ›
Gene Symbol:
5830434F19Rik
Gene Name:
RIKEN cDNA 5830434F19 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03941774_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 5830434F19Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK017969.1 - 1 749 95

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream mitogen-activated protein kinase associated protein 1 Mapkap1 AI591529,
D230039K05Rik,
Sin1
Sense Chr. 2 - 34406771 - 34624958 on NCBI Build GRCm38
Downstream GTPase activating protein and VPS9 domains 1 Gapvd1 2010005B09Rik,
4432404J10Rik,
AW108497,
Gapex-5,
RAP6,
RME-6,
mKIAA1521
258.9kb Anti-sense Chr. 2 - 34676179 - 34755263 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix ABI

Array Name Probe IDs Targets Target Type
Whole Mouse Genome G4122A A_52_P906992 AK017969.1 mouse_transcriptome
Agilent Six_ChIP_01_of_03_v2 A_68_P21127437 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21127448 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21127482 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21127484 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21127494 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21127455 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01995951 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01478761 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21127458 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21127501 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21127467 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21127480 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21127436 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21127468 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21127450 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21127432 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02268676 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21127439 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21127444 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01910556 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21127466 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01478771 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02221402 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21127486 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02221405 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21127472 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21127446 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21127496 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21127462 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21127478 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01416769 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21127487 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01995964 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21127440 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21127489 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21127488 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21127456 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01501066 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21127464 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02268679 - -
Array Name Probe IDs Targets Target Type
Affymetrix HT MG-430 PM Array Plate 1433037_PM_at - -
Affymetrix HT MG-430 PM Array Plate 1454447_PM_at - -
Mouse Genome 430 2.0 Array 1433037_PM_at - -
Mouse Genome 430 2.0 Array 1454447_PM_at - -
Mouse Genome 430 2.0 Array 1454447_at - -
Mouse Genome 430 2.0 Array 1433037_at - -
Mouse Expression Set 430 1454447_PM_at - -
Mouse Expression Set 430 1433037_PM_at - -
Mouse Expression Set 430 1433037_at - -
Mouse Expression Set 430 1454447_at - -
Affymetrix HT Mouse Genome 430B Array 1433037_at - -
Affymetrix HT Mouse Genome 430B Array 1454447_at - -
Affymetrix HT Mouse Genome 430B Array 1433037_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1454447_PM_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 309584 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0