This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03941122_s1
See other 5730406E14RIK GE Assays ›
Gene Symbol:
5730406E14Rik
Gene Name:
RIKEN cDNA 5730406E14 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03941122_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 5730406E14Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK017501.1 - 1 1146 81

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream protein tyrosine phosphatase,
receptor type,
N polypeptide 2
Ptprn2 4930425H11Rik,
IA2beta,
PTP-NP,
Phol,
mKIAA0387,
phogrin
Sense Chr. 12 - 116485692 - 117278167 on NCBI Build GRCm38
Downstream predicted gene 10421 Gm10421 ENSMUSG00000072792 111.5kb Anti-sense Chr. 12 - 117144523 - 117151269 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix HT MG-430 PM Array Plate 1433030_PM_at - -
Affymetrix HT MG-430 PM Array Plate 1454444_PM_at - -
Mouse Genome 430 2.0 Array 1454444_PM_at - -
Mouse Genome 430 2.0 Array 1454444_at - -
Mouse Genome 430 2.0 Array 1433030_PM_at - -
Mouse Genome 430 2.0 Array 1433030_at - -
Mouse Expression Set 430 1454444_PM_at - -
Mouse Expression Set 430 1454444_at - -
Mouse Expression Set 430 1433030_at - -
Mouse Expression Set 430 1433030_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1454444_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1454444_at - -
Affymetrix HT Mouse Genome 430B Array 1433030_at - -
Affymetrix HT Mouse Genome 430B Array 1433030_PM_at - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0