This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03941010_s1
See other DUSP8 GE Assays ›
Gene Symbol:
Dusp8
Gene Name:
dual specificity phosphatase 8
Gene Aliases:
5530400B01Rik, AI593498, Nttp1
Chromosome Location:
Chr.7: 142079487 - 142095880 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03941010_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dusp8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_008748.3 NP_032774.1 7 4650 65
GenBank mRNA AI849774.1 - 1 65
AK017419.1 - 1 1041 65
BQ043453.1 - 1 413 65
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BI078852.1 - 1 193 Chr. 7 - 142079486 - 142079828 [-] on Build GRCm38 65
GenBankmRNA AI850447.1 - 1 Chr. 7 - 142079501 - 142079752 [+] on Build GRCm38 65
GenBankmRNA CD742020.1 - 1 Chr. 7 - 142079494 - 142080020 [+] on Build GRCm38 65
GenBankmRNA BE688323.1 - 1 Chr. 7 - 142079496 - 142079861 [+] on Build GRCm38 65
GenBankmRNA BP772384.1 - 1 Chr. 7 - 142079494 - 142080036 [+] on Build GRCm38 65
GenBankmRNA AI852772.1 - 1 Chr. 7 - 142079494 - 142079854 [+] on Build GRCm38 65
GenBankmRNA AA103520.1 - 1 329 Chr. 7 - 142079586 - 142079983 [-] on Build GRCm38 65
GenBankmRNA BB802273.1 - 1 262 Chr. 7 - 142079506 - 142079917 [-] on Build GRCm38 65
GenBankmRNA AI843472.1 - 1 Chr. 7 - 142079494 - 142079827 [+] on Build GRCm38 65
GenBankmRNA CJ294565.1 - 1 360 Chr. 7 - 142079494 - 142080014 [-] on Build GRCm38 65
GenBankmRNA BX639338.1 - 1 Chr. 7 - 142079490 - 142079831 [+] on Build GRCm38 65
GenBankmRNA BB808699.1 - 1 274 Chr. 7 - 142079506 - 142079929 [-] on Build GRCm38 65
GenBankmRNA AW742686.1 - 1 Chr. 7 - 142079494 - 142080027 [+] on Build GRCm38 65
GenBankmRNA AI849760.1 - 1 Chr. 7 - 142079494 - 142079850 [+] on Build GRCm38 65
GenBankmRNA AI834944.1 - 1 Chr. 7 - 142079494 - 142079893 [+] on Build GRCm38 65
GenBankmRNA BU848181.1 - 236 Chr. - - [-] on Build 65
GenBankmRNA BE996276.1 - 1 Chr. 7 - 142079494 - 142079831 [+] on Build GRCm38 65

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream MOB kinase activator 2 Mob2 1110017M21Rik,
2700078K21Rik,
AI256456,
Mmh
-18.5kb Sense Chr. 7 - 142008535 - 142061034 on NCBI Build GRCm38
Downstream predicted gene 4553 Gm4553 82.3kb Sense Chr. 7 - 142163028 - 142167022 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Protein Phosphatase Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Dusp8

Panther Classification:

Molecular Function -

protein phosphatase

Gene Ontology Categories:

Function(s) Process(es)

protein dephosphorylation
dephosphorylation
phosphoprotein phosphatase activity
protein tyrosine phosphatase activity
protein tyrosine/serine/threonine phosphatase activity
hydrolase activity
phosphatase activity
MAP kinase tyrosine/serine/threonine phosphatase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0