This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03940873_s1
See other 5430425E15RIK GE Assays ›
Gene Symbol:
5430425E15Rik
Gene Name:
RIKEN cDNA 5430425E15 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03940873_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 5430425E15Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK017335.1 - 1 1045 63

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream predicted gene 10248 Gm10248 ENSMUSG00000068680 -425.1kb Anti-sense Chr. 14 - 23038194 - 23094571 on NCBI Build GRCm38
Downstream microRNA 7210 Mir7210 mir-7210,
mmu-mir-7210
568.0kb Sense Chr. 14 - 24088877 - 24088930 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix HT MG-430 PM Array Plate 1454515_PM_at - -
Affymetrix HT MG-430 PM Array Plate 1433233_PM_at - -
Mouse Genome 430 2.0 Array 1454515_at - -
Mouse Genome 430 2.0 Array 1433233_at - -
Mouse Genome 430 2.0 Array 1454515_PM_at - -
Mouse Genome 430 2.0 Array 1433233_PM_at - -
Mouse Expression Set 430 1433233_PM_at - -
Mouse Expression Set 430 1433233_at - -
Mouse Expression Set 430 1454515_PM_at - -
Mouse Expression Set 430 1454515_at - -
Affymetrix HT Mouse Genome 430B Array 1454515_at - -
Affymetrix HT Mouse Genome 430B Array 1454515_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1433233_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1433233_at - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0