This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03940754_m1
See other 5430401H09RIK GE Assays ›
Gene Symbol:
5430401H09Rik
Gene Name:
RIKEN cDNA 5430401H09 gene
Gene Aliases:
AW987716
Chromosome Location:
Chr.12: 32096593 - 32124760 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03940754_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 5430401H09Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK017255.1 - 1 - 2 83 74

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream protein kinase,
cAMP dependent regulatory,
type II beta
Prkar2b AI451071,
AW061005,
PKARIIbeta,
Pkarb2,
RII(beta)
-35.3kb Sense Chr. 12 - 31958450 - 32061343 on NCBI Build GRCm38
Downstream phosphoinositide-3-kinase,
catalytic,
gamma polypeptide
Pik3cg 5830428L06Rik,
PI3Kgamma,
p110gamma
49.4kb Sense Chr. 12 - 32173129 - 32208677 on NCBI Build GRCm38

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Mus musculus 1700 Probe spans exons

MicroArray Probes:

ABI

Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 450832 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0