This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03940680_s1
See other 5033430J17RIK GE Assays ›
Gene Symbol:
5033430J17Rik
Gene Name:
RIKEN cDNA 5033430J17 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03940680_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 5033430J17Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK017208.1 - 1 1050 85

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream PDZ domain containing 2 Pdzd2 4930537L06Rik,
A930022H17Rik,
Gm82,
Pdzk3,
mKIAA0300
Sense Chr. 15 - 12357054 - 12739973 on NCBI Build GRCm38
Downstream RIKEN cDNA 6030458C11 gene 6030458C11Rik 271.4kb Sense Chr. 15 - 12808177 - 12824688 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P156694 - -
Array Name Probe IDs Targets Target Type
Affymetrix HT MG-430 PM Array Plate 1433207_PM_at - -
Affymetrix HT MG-430 PM Array Plate 1433206_PM_at - -
Mouse Genome 430 2.0 Array 1433206_at - -
Mouse Genome 430 2.0 Array 1433206_PM_at - -
Mouse Genome 430 2.0 Array 1433207_PM_at - -
Mouse Genome 430 2.0 Array 1433207_at - -
Mouse Expression Set 430 1433206_PM_at - -
Mouse Expression Set 430 1433207_at - -
Mouse Expression Set 430 1433206_at - -
Mouse Expression Set 430 1433207_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1433207_at - -
Affymetrix HT Mouse Genome 430B Array 1433206_at - -
Affymetrix HT Mouse Genome 430B Array 1433206_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1433207_PM_at - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 101090176 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0