This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03940399_s1
See other 4933428L12RIK GE Assays ›
Gene Symbol:
4933428L12Rik
Gene Name:
RIKEN cDNA 4933428L12 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03940399_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 4933428L12Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK016971.1 - 1 945 70
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA EL962590.1 - 1 162 Chr. 14 - 24447340 - 24447904 [+] on Build GRCm38 70

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream predicted gene,
41109
Gm41109 -16.7kb Anti-sense Chr. 14 - 24409976 - 24429854 on NCBI Build GRCm38
Downstream polymerase (RNA) III (DNA directed) polypeptide A Polr3a 9330175N20Rik,
BC053071,
RPC1,
RPC155
766.0 Anti-sense Chr. 14 - 24448694 - 24487186 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix ABI

Array Name Probe IDs Targets Target Type
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 533757 - -
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106457 - -
Array Name Probe IDs Targets Target Type
Affymetrix HT MG-430 PM Array Plate 1433099_PM_at - -
Affymetrix HT MG-430 PM Array Plate 1454476_PM_at - -
Mouse Genome 430 2.0 Array 1433099_PM_at - -
Mouse Genome 430 2.0 Array 1454476_PM_at - -
Mouse Genome 430 2.0 Array 1454476_at - -
Mouse Genome 430 2.0 Array 1433099_at - -
Mouse Expression Set 430 1454476_at - -
Mouse Expression Set 430 1454476_PM_at - -
Mouse Expression Set 430 1433099_at - -
Mouse Expression Set 430 1433099_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1433099_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1433099_at - -
Affymetrix HT Mouse Genome 430B Array 1454476_at - -
Affymetrix HT Mouse Genome 430B Array 1454476_PM_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 748969 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0