Assay Details
| Gene Symbol: 4933428L01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK016970.1 | - | 1 | 544 | 93 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ER membrane protein complex subunit 2 | Emc2 |
4921531G14Rik, AV060620, AW209495, Ttc35 |
-171.8kb | Anti-sense | Chr. 15 - 43477214 - 43527777 on NCBI Build GRCm38 |
| Downstream | transmembrane protein 74 | Tmem74 |
AA549547, B230382K22Rik |
165.7kb | Sense | Chr. 15 - 43866695 - 43870612 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4933428L01Rik
Genomic Map