This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03940272_s1
See other 4933424C08RIK GE Assays ›
Gene Symbol:
4933424C08Rik
Gene Name:
RIKEN cDNA 4933424C08 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03940272_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 4933424C08Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK016884.1 - 1 851 98
AK048826.1 - 1 972 98
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BF721876.1 - 1 Chr. 4 - 126968718 - 126969305 [-] on Build GRCm38 98
GenBankmRNA BB391148.2 - 1 362 Chr. 4 - 126968743 - 126969379 [+] on Build GRCm38 98
GenBankmRNA BE646755.1 - 1 Chr. 4 - 126968773 - 126969214 [-] on Build GRCm38 98

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream expressed sequence AU040320 AU040320 A730047D20Rik,
Kiaa0319l
-98.6kb Sense Chr. 4 - 126753555 - 126869694 on NCBI Build GRCm38
Downstream splicing factor proline/glutamine rich (polypyrimidine tract binding protein associated) Sfpq 1110004P21Rik,
2810416M14Rik,
5730453G22Rik,
9030402K04Rik,
AU021830,
D4Ertd314e,
Gm12940,
OTTMUSG00000009329,
PSF,
REP1
51.2kb Sense Chr. 4 - 127020560 - 127037016 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 529694 - -
Array Name Probe IDs Targets Target Type
Affymetrix HT MG-430 PM Array Plate 1429867_PM_at - -
Mouse Genome 430 2.0 Array 1429867_at - -
Mouse Genome 430 2.0 Array 1429867_PM_at - -
Mouse Expression Set 430 1429867_at - -
Mouse Expression Set 430 1429867_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1429867_at - -
Affymetrix HT Mouse Genome 430B Array 1429867_PM_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 418977 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 102480440 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header