This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03940263_s1
See other 4933423N12RIK GE Assays ›
Gene Symbol:
4933423N12Rik
Gene Name:
RIKEN cDNA 4933423N12 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03940263_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 4933423N12Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK016878.1 - 1 745 98

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream asparagine-linked glycosylation 10B (alpha-1,2-glucosyltransferase) Alg10b AA469671,
AU045404,
AW123895,
C85569,
Deaf1,
Gm917,
nse5
-115.4kb Anti-sense Chr. 15 - 90224311 - 90230554 on NCBI Build GRCm38
Downstream predicted gene,
36480
Gm36480 6.9kb Anti-sense Chr. 15 - 90353710 - 90368444 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

MicroArray Probes:

Agilent ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Six_ChIP_03_of_03_v2 A_68_P30429623 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30429596 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30429616 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30429581 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30429589 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30429579 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30429588 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30429622 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30429606 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30429629 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30429594 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30429580 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30429593 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30429624 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30429598 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30429586 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30429582 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30429608 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30429612 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30429591 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12176422 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30429603 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30429618 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30429600 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30429577 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30429621 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30429602 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30429584 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30429611 - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 627291 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 105050112 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0