This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03940166_s1
See other 4933416A02RIK GE Assays ›
Gene Symbol:
4933416A02Rik
Gene Name:
RIKEN cDNA 4933416A02 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03940166_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 4933416A02Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK016828.1 - 1 143 104
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY716502.1 - 1 143 Chr. 15 - 84580517 - 84581441 [-] on Build GRCm38 104

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream leucine zipper,
down-regulated in cancer 1-like
Ldoc1l BC058638,
Mar6,
Mart6
-21.7kb Sense Chr. 15 - 84553398 - 84557823 on NCBI Build GRCm38
Downstream proline rich 5 (renal) Prr5 AU043908,
Arhgap8,
C030017C09Rik,
C78947,
Protor-1,
Protor1
81.7kb Anti-sense Chr. 15 - 84663171 - 84706409 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix ABI

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2250514 AK016828.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 634458 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30395654 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12453136 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30395591 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30395651 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30395586 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30395655 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30395659 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30395622 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30395620 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30395617 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30395661 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30395624 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12179044 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30395590 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30395588 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30395671 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12343985 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P11956270 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12305305 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12343987 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30395584 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30395594 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12179036 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30395627 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P11956267 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30395658 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30395663 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30395593 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30395589 - -
Array Name Probe IDs Targets Target Type
Affymetrix HT MG-430 PM Array Plate 1432925_PM_at - -
Affymetrix HT MG-430 PM Array Plate 1460530_PM_at - -
Mouse Genome 430 2.0 Array 1460530_at - -
Mouse Genome 430 2.0 Array 1432925_at - -
Mouse Genome 430 2.0 Array 1460530_PM_at - -
Mouse Genome 430 2.0 Array 1432925_PM_at - -
Mouse Expression Set 430 1432925_PM_at - -
Mouse Expression Set 430 1432925_at - -
Mouse Expression Set 430 1460530_PM_at - -
Mouse Expression Set 430 1460530_at - -
Affymetrix HT Mouse Genome 430B Array 1432925_at - -
Affymetrix HT Mouse Genome 430B Array 1432925_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1460530_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1460530_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 317663 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header