This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03940134_s1
See other 4933413I22RIK GE Assays ›
Gene Symbol:
4933413I22Rik
Gene Name:
RIKEN cDNA 4933413I22 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03940134_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 4933413I22Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK016806.1 - 1 1250 103
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AV280310.1 - 1 183 Chr. 8 - 11726065 - 11726309 [-] on Build GRCm38 103

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream RIKEN cDNA 1700128E19 gene 1700128E19Rik -5.5kb Anti-sense Chr. 8 - 11713260 - 11720542 on NCBI Build GRCm38
Downstream Rho guanine nucleotide exchange factor (GEF7) Arhgef7 Cool,
PIX,
Pak3bp,
betaPix,
betaPix-b,
betaPix-c,
cool-1,
mKIAA0142,
p85Cool1,
p85SPR
730.0 Anti-sense Chr. 8 - 11728105 - 11835219 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix

Array Name Probe IDs Targets Target Type
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 863095 - -
Whole Mouse Genome G4122A A_52_P1098803 AK016806.1 mouse_transcriptome
Array Name Probe IDs Targets Target Type
Affymetrix HT MG-430 PM Array Plate 1432594_PM_at - -
Affymetrix HT MG-430 PM Array Plate 1432595_PM_at - -
Mouse Genome 430 2.0 Array 1432595_at - -
Mouse Genome 430 2.0 Array 1432594_at - -
Mouse Genome 430 2.0 Array 1432595_PM_at - -
Mouse Genome 430 2.0 Array 1432594_PM_at - -
Mouse Expression Set 430 1432595_PM_at - -
Mouse Expression Set 430 1432594_at - -
Mouse Expression Set 430 1432594_PM_at - -
Mouse Expression Set 430 1432595_at - -
Affymetrix HT Mouse Genome 430B Array 1432595_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1432595_at - -
Affymetrix HT Mouse Genome 430B Array 1432594_at - -
Affymetrix HT Mouse Genome 430B Array 1432594_PM_at - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header