This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03940102_s1
See other 4933411K05RIK GE Assays ›
Gene Symbol:
4933411K05Rik
Gene Name:
RIKEN cDNA 4933411K05 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03940102_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 4933411K05Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK016774.1 - 1 1152 81

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream centrosomal protein 63 Cep63 4921501M07,
AL450317.13gm1,
AW107703,
CD20R,
D9Mgc41,
D9Mgc48e,
ET2
-2.2kb Anti-sense Chr. 9 - 102586578 - 102626353 on NCBI Build GRCm38
Downstream angiomotin-like 2 Amotl2 AW549739,
C79691,
Lccp,
MASCOT,
mKIAA0989
87.6kb Sense Chr. 9 - 102717294 - 102733418 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix HT MG-430 PM Array Plate 1430270_PM_at - -
Mouse Genome 430 2.0 Array 1430270_PM_at - -
Mouse Genome 430 2.0 Array 1430270_at - -
Mouse Expression Set 430 1430270_PM_at - -
Mouse Expression Set 430 1430270_at - -
Affymetrix HT Mouse Genome 430B Array 1430270_at - -
Affymetrix HT Mouse Genome 430B Array 1430270_PM_at - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header