This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03939464_s1
See other 4930557B21RIK GE Assays ›
Gene Symbol:
4930557B21Rik
Gene Name:
RIKEN cDNA 4930557B21 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03939464_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 4930557B21Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK016159.1 - 1 452 98
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY715762.1 - 1 453 Chr. 19 - 45080006 - 45080583 [+] on Build GRCm38 98

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream Kazal-type serine peptidase inhibitor domain 1 Kazald1 AI842353,
Bono1,
IGFBP-rP10
-719.0 Sense Chr. 19 - 45074485 - 45079289 on NCBI Build GRCm38
Downstream T cell leukemia,
homeobox 1
Tlx1 Hox-11,
Hox11
69.5kb Sense Chr. 19 - 45150079 - 45156943 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix ABI

Array Name Probe IDs Targets Target Type
Whole Mouse Genome G4122A A_52_P1003306 AK016159.1 mouse_transcriptome
Array Name Probe IDs Targets Target Type
Affymetrix HT MG-430 PM Array Plate 1432822_PM_at - -
Affymetrix HT MG-430 PM Array Plate 1454370_PM_at - -
Mouse Genome 430 2.0 Array 1454370_at - -
Mouse Genome 430 2.0 Array 1432822_at - -
Mouse Genome 430 2.0 Array 1432822_PM_at - -
Mouse Genome 430 2.0 Array 1454370_PM_at - -
Mouse Expression Set 430 1432822_PM_at - -
Mouse Expression Set 430 1432822_at - -
Mouse Expression Set 430 1454370_PM_at - -
Mouse Expression Set 430 1454370_at - -
Affymetrix HT Mouse Genome 430B Array 1432822_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1432822_at - -
Affymetrix HT Mouse Genome 430B Array 1454370_at - -
Affymetrix HT Mouse Genome 430B Array 1454370_PM_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 837662 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header