This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03939428_s1
See other 4930554P06RIK GE Assays ›
Gene Symbol:
4930554P06Rik
Gene Name:
RIKEN cDNA 4930554P06 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03939428_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 4930554P06Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK016124.1 - 1 849 71

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream aminoadipate-semialdehyde synthase Aass LKR,
LKR/SDH,
LOR,
LOR/SDH,
Lorsdh,
SDH
-47.5kb Sense Chr. 6 - 23072173 - 23132986 on NCBI Build GRCm38
Downstream Fez family zinc finger 1 Fezf1 3110069A13Rik,
Fez,
Zfp312-like,
fez-like
63.6kb Sense Chr. 6 - 23245044 - 23255368 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2259889 AK016124.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 22939 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 17831 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05834026 - -
Whole Mouse Genome G4122A A_51_P360877 AK016124.1 mouse_transcriptome
Agilent Six_ChIP_02_of_03_v2 A_68_P24295001 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24295068 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24295051 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24295063 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24294995 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24295065 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24295003 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24295066 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24295069 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24295058 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24295050 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24294988 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24295053 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24295048 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24294999 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05636440 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24295049 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24295055 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05618944 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24295071 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24294997 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24295059 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24295006 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24294991 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24294989 - -
Array Name Probe IDs Targets Target Type
Affymetrix HT MG-430 PM Array Plate 1454365_PM_at - -
Affymetrix HT MG-430 PM Array Plate 1432812_PM_at - -
Mouse Genome 430 2.0 Array 1454365_PM_at - -
Mouse Genome 430 2.0 Array 1432812_at - -
Mouse Genome 430 2.0 Array 1432812_PM_at - -
Mouse Genome 430 2.0 Array 1454365_at - -
Mouse Expression Set 430 1454365_PM_at - -
Mouse Expression Set 430 1432812_PM_at - -
Mouse Expression Set 430 1454365_at - -
Mouse Expression Set 430 1432812_at - -
Affymetrix HT Mouse Genome 430B Array 1432812_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1432812_at - -
Affymetrix HT Mouse Genome 430B Array 1454365_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1454365_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 866901 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 104230592 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0