This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03939254_s1
See other 4930535F04RIK GE Assays ›
Gene Symbol:
4930535F04Rik
Gene Name:
RIKEN cDNA 4930535F04 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03939254_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 4930535F04Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK015981.1 - 1 549 90
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY715551.1 - 1 547 Chr. 19 - 48991019 - 48991969 [-] on Build GRCm38 90

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream sortilin-related VPS10 domain containing receptor 3 Sorcs3 6330404A12Rik,
AW045579
-185.0kb Anti-sense Chr. 19 - 48206025 - 48805505 on NCBI Build GRCm38
Downstream sortilin-related VPS10 domain containing receptor 1 Sorcs1 Sorcs,
mSorCS
1.2mb Sense Chr. 19 - 50143301 - 50679136 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Whole Mouse Genome G4122A A_51_P315911 AK015981.1 mouse_transcriptome
Agilent Six_ChIP_03_of_03_v2 A_68_P32159282 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32159283 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32159278 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32159286 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32159288 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32159290 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14349837 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32159287 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32159279 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14349838 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14484460 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32159297 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32159285 - -
Array Name Probe IDs Targets Target Type
Affymetrix HT MG-430 PM Array Plate 1433045_PM_at - -
Affymetrix HT MG-430 PM Array Plate 1433046_PM_at - -
Mouse Genome 430 2.0 Array 1433046_PM_at - -
Mouse Genome 430 2.0 Array 1433046_at - -
Mouse Genome 430 2.0 Array 1433045_at - -
Mouse Genome 430 2.0 Array 1433045_PM_at - -
Mouse Expression Set 430 1433045_at - -
Mouse Expression Set 430 1433046_at - -
Mouse Expression Set 430 1433045_PM_at - -
Mouse Expression Set 430 1433046_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1433046_at - -
Affymetrix HT Mouse Genome 430B Array 1433045_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1433045_at - -
Affymetrix HT Mouse Genome 430B Array 1433046_PM_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 433282 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 106760044 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0