This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03939185_s1
See other 4930526M16RIK GE Assays ›
Gene Symbol:
4930526M16Rik
Gene Name:
RIKEN cDNA 4930526M16 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03939185_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 4930526M16Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK015908.1 - 1 839 81

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream RIKEN cDNA 1700027F09 gene 1700027F09Rik -116.0 Anti-sense Chr. 5 - 64454813 - 64467942 on NCBI Build GRCm38
Downstream RIKEN cDNA C030018K13 gene C030018K13Rik 8.0kb Sense Chr. 5 - 64477008 - 64483967 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

MicroArray Probes:

Agilent

Array Name Probe IDs Targets Target Type
Agilent Six_ChIP_01_of_03_v2 A_68_P23751286 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23751289 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04949206 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23751282 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04181092 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0