This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03939096_s1
See other 4930517G19RIK GE Assays ›
Gene Symbol:
4930517G19Rik
Gene Name:
RIKEN cDNA 4930517G19 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03939096_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 4930517G19Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK015812.1 - 1 1451 84

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream solute carrier family 6 (neurotransmitter transporter,
taurine),
member 6
Slc6a6 AA589629,
C80501,
Taut
Anti-sense Chr. 6 - 91684044 - 91759063 on NCBI Build GRCm38
Downstream glutamate receptor interacting protein 2 Grip2 69.9kb Sense Chr. 6 - 91761508 - 91843954 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2421835 AK015812.1 mouse_transcriptome
Array Name Probe IDs Targets Target Type
Affymetrix HT MG-430 PM Array Plate 1432692_PM_at - -
Affymetrix HT MG-430 PM Array Plate 1432693_PM_at - -
Mouse Genome 430 2.0 Array 1432692_at - -
Mouse Genome 430 2.0 Array 1432693_PM_at - -
Mouse Genome 430 2.0 Array 1432693_at - -
Mouse Genome 430 2.0 Array 1432692_PM_at - -
Mouse Expression Set 430 1432693_at - -
Mouse Expression Set 430 1432692_PM_at - -
Mouse Expression Set 430 1432692_at - -
Mouse Expression Set 430 1432693_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1432692_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1432693_at - -
Affymetrix HT Mouse Genome 430B Array 1432692_at - -
Affymetrix HT Mouse Genome 430B Array 1432693_PM_at - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0