This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03938890_s1
See other 4930478M09RIK GE Assays ›
Gene Symbol:
4930478M09Rik
Gene Name:
RIKEN cDNA 4930478M09 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03938890_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 4930478M09Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK015588.1 - 1 348 77
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY715138.1 - 1 348 Chr. 5 - 31451513 - 31452331 [-] on Build GRCm38 77

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream glucokinase regulatory protein Gckr GKRP -123.9kb Anti-sense Chr. 5 - 31297254 - 31327302 on NCBI Build GRCm38
Downstream zinc finger protein 512 Zfp512 2500002M11Rik,
AI450173,
D230008H22,
Znf512
85.0 Anti-sense Chr. 5 - 31452416 - 31481754 on NCBI Build GRCm38

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 855920 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04356919 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23578076 - -
Array Name Probe IDs Targets Target Type
Affymetrix HT MG-430 PM Array Plate 1433301_PM_at - -
Affymetrix HT MG-430 PM Array Plate 1454550_PM_at - -
Mouse Genome 430 2.0 Array 1454550_at - -
Mouse Genome 430 2.0 Array 1433301_at - -
Mouse Genome 430 2.0 Array 1454550_PM_at - -
Mouse Genome 430 2.0 Array 1433301_PM_at - -
Mouse Expression Set 430 1433301_PM_at - -
Mouse Expression Set 430 1454550_PM_at - -
Mouse Expression Set 430 1433301_at - -
Mouse Expression Set 430 1454550_at - -
Affymetrix HT Mouse Genome 430B Array 1433301_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1454550_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1433301_at - -
Affymetrix HT Mouse Genome 430B Array 1454550_at - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 106450402 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header