This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03938824_s1
See other 4930466K18RIK GE Assays ›
Gene Symbol:
4930466K18Rik
Gene Name:
RIKEN cDNA 4930466K18 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03938824_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 4930466K18Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK015515.1 - 1 545 104
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AU067642.1 - 1 Chr. 10 - 53614485 - 53615277 [-] on Build GRCm38 104
GenBankmRNA AU067685.1 - 1 Chr. 10 - 53614539 - 53615277 [-] on Build GRCm38 104
GenBankmRNA BU610016.1 - 1 629 Chr. 10 - 53614284 - 53614999 [+] on Build GRCm38 104

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream minichromosome maintenance complex component 9 Mcm9 9030408O17Rik,
Gm235,
Mcmdc1
Anti-sense Chr. 10 - 53536315 - 53630439 on NCBI Build GRCm38
Downstream family with sequence similarity 184,
member A
Fam184a 3110012E06Rik,
4930438C08Rik,
4930589M24Rik
17.9kb Anti-sense Chr. 10 - 53633128 - 53751231 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix HT MG-430 PM Array Plate 1439671_PM_at - -
Mouse Genome 430 2.0 Array 1439671_PM_at - -
Mouse Genome 430 2.0 Array 1439671_at - -
Mouse Expression Set 430 1439671_PM_at - -
Mouse Expression Set 430 1439671_at - -
Affymetrix HT Mouse Genome 430B Array 1439671_at - -
Affymetrix HT Mouse Genome 430B Array 1439671_PM_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 858467 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0