This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03938764_s1
See other 4930456A14RIK GE Assays ›
Gene Symbol:
4930456A14Rik
Gene Name:
RIKEN cDNA 4930456A14 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03938764_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 4930456A14Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK015472.1 - 1 1350 80
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA CK343182.1 - 1 Chr. 7 - 126504791 - 126505466 [-] on Build GRCm38 80

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream ataxin 2-like Atxn2l A2D,
A2LG,
A2RP,
A2lp,
Atxnl2
-96.0 Anti-sense Chr. 7 - 126491708 - 126503644 on NCBI Build GRCm38
Downstream eukaryotic translation initiation factor 3,
subunit C
Eif3c 110kDa,
3230401O13Rik,
Eif3s8,
NIPIL(A3),
NipilA3,
Xs,
Xsl
41.4kb Anti-sense Chr. 7 - 126546911 - 126566366 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix ABI

Array Name Probe IDs Targets Target Type
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 439486 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 99926 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06359144 - -
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P132799 - -
Array Name Probe IDs Targets Target Type
Affymetrix HT MG-430 PM Array Plate 1432968_PM_at - -
Affymetrix HT MG-430 PM Array Plate 1432967_PM_at - -
Mouse Genome 430 2.0 Array 1432968_PM_at - -
Mouse Genome 430 2.0 Array 1432968_at - -
Mouse Genome 430 2.0 Array 1432967_PM_at - -
Mouse Genome 430 2.0 Array 1432967_at - -
Mouse Expression Set 430 1432968_at - -
Mouse Expression Set 430 1432967_at - -
Mouse Expression Set 430 1432968_PM_at - -
Mouse Expression Set 430 1432967_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1432967_at - -
Affymetrix HT Mouse Genome 430B Array 1432967_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1432968_at - -
Affymetrix HT Mouse Genome 430B Array 1432968_PM_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 510039 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0