This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03937997_m1
See other D030025E07RIK GE Assays ›
Gene Symbol:
D030025E07Rik
Gene Name:
RIKEN cDNA D030025E07 gene
Gene Aliases:
-
Chromosome Location:
Chr.3: 128117015 - 128231605 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03937997_m1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: D030025E07Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK048251.1 - 1 - 2 234 74
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BB648889.1 - 1 - 2 234 Chr. 3 - 128149575 - 128231553 [-] on Build GRCm38 74

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream RIKEN cDNA 9830132P13 gene 9830132P13Rik 9830132P13 -230.9kb Anti-sense Chr. 3 - 127916171 - 127955220 on NCBI Build GRCm38
Downstream paired-like homeodomain transcription factor 2 Pitx2 9430085M16Rik,
Brx1,
Brx1b,
Munc30,
Otlx2,
Ptx2,
Rieg
968.3kb Anti-sense Chr. 3 - 129199878 - 129219594 on NCBI Build GRCm38

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mus musculus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0