This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03937578_s1
See other D430040D24RIK GE Assays ›
Gene Symbol:
D430040D24Rik
Gene Name:
RIKEN cDNA D430040D24 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03937578_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: D430040D24Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK047283.1 - 1 753 88
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA CJ161180.1 - 1 76 Chr. 1 - 36558377 - 36558766 [+] on Build GRCm38 88
GenBankmRNA BI693401.1 - 2 238 Chr. 1 - 36558135 - 36558899 [+] on Build GRCm38 89

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream sema domain,
immunoglobulin domain (Ig),
transmembrane domain (TM) and short cytoplasmic domain,
(semaphorin) 4C
Sema4c AI426163,
M-Sema F,
Semacl1,
Semaf,
Semai,
sema I
Anti-sense Chr. 1 - 36548640 - 36558381 on NCBI Build GRCm38
Downstream family with sequence similarity 178,
member B
Fam178b 1700024G10Rik,
Gm990
3.5kb Anti-sense Chr. 1 - 36562696 - 36683190 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0