This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03937099_s1
See other B230337E12RIK GE Assays ›
Gene Symbol:
B230337E12Rik
Gene Name:
RIKEN cDNA B230337E12 gene
Gene Aliases:
AI115432
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03937099_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: B230337E12Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AI846867.1 - 2 293 86
AK046039.1 - 1 1755 86
AK144670.1 - 1 2165 86
BE199656.1 - 1 86
BM120593.2 - 1 86
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA CJ261365.1 - 1 217 Chr. 1 - 119652469 - 119652928 [-] on Build GRCm38 86
GenBankmRNA BE447126.1 - 1 Chr. 1 - 119652474 - 119652917 [+] on Build GRCm38 86
GenBankmRNA CJ261572.1 - 1 214 Chr. 1 - 119652469 - 119652919 [-] on Build GRCm38 86
GenBankmRNA BB797479.1 - 1 167 Chr. 1 - 119652470 - 119652878 [-] on Build GRCm38 86
GenBankmRNA BQ557266.1 - 1 Chr. 1 - 119652469 - 119653103 [+] on Build GRCm38 86
GenBankmRNA BE993100.1 - 1 Chr. 1 - 119652467 - 119652814 [+] on Build GRCm38 86
GenBankmRNA BB800821.1 - 1 130 Chr. 1 - 119652470 - 119652841 [-] on Build GRCm38 86
GenBankmRNA BE860941.1 - 1 Chr. 1 - 119652537 - 119652918 [+] on Build GRCm38 86
GenBankmRNA BI106368.1 - 54 Chr. - - [-] on Build 87
GenBankmRNA BB312663.2 - 1 419 Chr. 1 - 119652470 - 119653127 [-] on Build GRCm38 86
GenBankmRNA BM933721.1 - 1 117 Chr. 1 - 119652467 - 119652814 [-] on Build GRCm38 86
GenBankmRNA BE849998.1 - 1 418 Chr. 1 - 119652648 - 119653129 [-] on Build GRCm38 86
GenBankmRNA BP760575.1 - 1 Chr. 1 - 119652467 - 119653003 [+] on Build GRCm38 86
GenBankmRNA CA893651.1 - 1 Chr. 1 - 119652469 - 119652810 [+] on Build GRCm38 86
GenBankmRNA CA888411.1 - 1 Chr. 1 - 119652541 - 119653105 [+] on Build GRCm38 86
GenBankmRNA BM938915.1 - 1 117 Chr. 1 - 119652467 - 119652814 [-] on Build GRCm38 86
GenBankmRNA BF018019.1 - 1 Chr. 1 - 119652472 - 119652955 [+] on Build GRCm38 86
GenBankmRNA BE849977.1 - 1 418 Chr. 1 - 119652650 - 119653129 [-] on Build GRCm38 86
GenBankmRNA BF152728.1 - 1 Chr. 1 - 119652467 - 119652792 [+] on Build GRCm38 86

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream protein tyrosine phosphatase,
non-receptor type 4
Ptpn4 PTPMEG,
Ptn4,
TEP,
TEP/mPTPMEG,
hPTP-MEG
Sense Chr. 1 - 119652467 - 119837542 on NCBI Build GRCm38
Downstream transmembrane protein 177 Tmem177 2300008B03Rik 253.4kb Sense Chr. 1 - 119907890 - 119913168 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 813136 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P162835 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P162835 - -
Whole Mouse Genome G4122A A_52_P402960 AI846867.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P402960 AK046039.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P402960 AK144670.1 mouse_transcriptome
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P162835 - -
Array Name Probe IDs Targets Target Type
Affymetrix HT MG-430 PM Array Plate 1434671_PM_at - -
Mouse Genome 430 2.0 Array 1434671_at - -
Mouse Genome 430 2.0 Array 1434671_PM_at - -
Mouse Expression Set 430 1434671_at - -
Mouse Expression Set 430 1434671_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1434671_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1434671_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 457339 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 101170593 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0