This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03936956_s1
See other B230114P17RIK GE Assays ›
Gene Symbol:
B230114P17Rik
Gene Name:
RIKEN cDNA B230114P17 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03936956_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: B230114P17Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK045415.1 - 1 2149 68
BB304874.2 - 1 434 68

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream protein tyrosine phosphatase,
receptor type,
f polypeptide (PTPRF),
interacting protein (liprin),
alpha 2
Ppfia2 5330438O12,
B230207K17Rik,
E130120L08Rik,
mKIAA4112
-49.0 Anti-sense Chr. 10 - 106468872 - 106933468 on NCBI Build GRCm38
Downstream acyl-CoA synthetase short-chain family member 3 Acss3 8430416H19Rik,
Gm874
212.0 Sense Chr. 10 - 106936164 - 107123664 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix ABI

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2340448 AK045415.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2340448 BB284517.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2340448 BB304874.2 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 325119 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 174474 - -
Whole Mouse Genome G4122A A_51_P424954 AK045415.1 mouse_transcriptome
Array Name Probe IDs Targets Target Type
Affymetrix HT MG-430 PM Array Plate 1456295_PM_at - -
Mouse Genome 430 2.0 Array 1456295_at - -
Mouse Genome 430 2.0 Array 1456295_PM_at - -
Mouse Expression Set 430 1456295_PM_at - -
Mouse Expression Set 430 1456295_at - -
Affymetrix HT Mouse Genome 430B Array 1456295_at - -
Affymetrix HT Mouse Genome 430B Array 1456295_PM_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 539613 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0