This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03936659_s1
See other C730002L08RIK GE Assays ›
Gene Symbol:
C730002L08Rik
Gene Name:
RIKEN cDNA C730002L08 gene
Gene Aliases:
-
Chromosome Location:
Chr.19: 20534527 - 20552330 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03936659_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: C730002L08Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_045778.1 3 1911 82
GenBank mRNA AK043892.1 - 1 1143 82
AK050017.1 - 3 1913 82

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream RIKEN cDNA E030003E18 gene E030003E18Rik 1700127G21Rik Anti-sense Chr. 19 - 20492715 - 20556410 on NCBI Build GRCm38
Downstream aldehyde dehydrogenase family 1,
subfamily A1
Aldh1a1 Ahd-2,
Ahd2,
Aldh1,
Aldh1a2,
E1,
Raldh1
65.3kb Anti-sense Chr. 19 - 20601982 - 20643462 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2324182 AK050017.1 mouse_transcriptome
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P146321 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2324182 AK050017.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P577315 AK043892.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P577315 AK050017.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129576 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32001470 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14292762 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32001478 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32001475 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32001485 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32001458 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14459802 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32001450 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32001486 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14459799 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32001464 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32001447 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32001449 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32001490 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32001489 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32001472 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14305858 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32001483 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32001460 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32001461 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32001484 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32001476 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32001474 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32001453 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14459808 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1900011245.mm.1 NR_045778.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900011245.mm.1 NR_045778.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900011245.mm.1 TC1900001243.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 408749 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 106420035 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0