This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03936053_s1
See other FAM222B GE Assays ›
Gene Symbol:
Fam222b
Gene Name:
family with sequence similarity 222, member B
Gene Aliases:
9630020I17Rik
Chromosome Location:
Chr.11: 78094666 - 78157339 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03936053_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Fam222b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK041269.1 - 1 144 80
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BB226091.2 - 1 183 Chr. 11 - 78097912 - 78098397 [+] on Build GRCm38 80
GenBankmRNA BB637765.1 - 1 144 Chr. 11 - 78097913 - 78098409 [+] on Build GRCm38 80

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream Era (G-protein)-like 1 (E. coli) Eral1 2610524P08Rik,
9130407C09Rik,
AU019798,
Era,
M-ERA,
MERA-S,
MERA-W
-17.5kb Anti-sense Chr. 11 - 78073376 - 78080383 on NCBI Build GRCm38
Downstream TNF receptor associated factor 4 Traf4 A530032M13Rik,
CART1,
msp2
60.0kb Anti-sense Chr. 11 - 78158423 - 78165569 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0