This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03935970_s1
See other 4632404H12RIK GE Assays ›
Gene Symbol:
4632404H12Rik
Gene Name:
RIKEN cDNA 4632404H12 gene
Gene Aliases:
-
Chromosome Location:
Chr.3: 89768430 - 89773233 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03935970_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 4632404H12Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK040741.1 - 1 1150 109
AK040882.1 - 1 1178 109
AK047674.1 - 1 1167 109
AK048101.1 - 1 1336 109
AK084338.1 - 1 1168 109
AK164170.1 - 1 1227 109
AK169469.1 - 1 1175 109
BC080683.1 - 1 1166 109
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BQ930208.1 - 1 349 Chr. 3 - 89771520 - 89772408 [-] on Build GRCm38 109
GenBankmRNA AI152939.1 - 1 277 Chr. 3 - 89771960 - 89772326 [-] on Build GRCm38 109

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream cholinergic receptor,
nicotinic,
beta polypeptide 2 (neuronal)
Chrnb2 Acrb-2,
Acrb2,
C030030P04Rik,
[b]2-nAchR
-6.8kb Sense Chr. 3 - 89753448 - 89764632 on NCBI Build GRCm38
Downstream ubiquitin-conjugating enzyme E2Q (putative) 1 Ube2q1 1110002C01Rik,
2310012M18Rik,
AI854014,
Gtap,
NICE-5,
PRO3094,
Ube2q
396.0 Anti-sense Chr. 3 - 89773609 - 89783997 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22321609 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22321611 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22321608 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P483483 AK014549.2 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P483483 AK014549.2 mouse_transcriptome
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P145531 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22321586 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22321603 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22321606 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22321584 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22321605 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22321595 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22321578 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22321588 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22321583 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22321611 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22321591 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22321601 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22321598 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22321608 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22321580 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22321609 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22321599 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22321594 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22321600 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22321577 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22321590 - -
Whole Mouse Genome G4122A A_52_P389863 AK164170.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P389863 AK169469.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P389863 AW045319.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P389863 BC063049.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P389863 AK048101.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P483483 AK014549.2 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P483483 AK014549.2 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P133135 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22321608 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22321609 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22321611 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10499653 - -
Affymetrix Mouse Gene 1.0 ST Array 10499654 - -
Affymetrix HT MG-430 PM Array Plate 1431820_PM_at - -
Affymetrix HT MG-430 PM Array Plate 1438209_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10499652 - -
Mouse Genome 430 2.0 Array 1431820_PM_at - -
Mouse Genome 430 2.0 Array 1438209_PM_at - -
Mouse Genome 430 2.0 Array 1431820_at - -
Mouse Genome 430 2.0 Array 1438209_at - -
Mouse Expression Set 430 1431820_at - -
Mouse Expression Set 430 1431820_PM_at - -
Mouse Expression Set 430 1438209_at - -
Mouse Expression Set 430 1438209_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1438209_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1431820_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1431820_at - -
Affymetrix HT Mouse Genome 430B Array 1438209_at - -
Array Name Probe IDs Targets Target Type
Illumina mouse-6 v1.0 expression beadchip 5900168 - -
Illumina mouse-6 v1.0 expression beadchip 5720484 - -
Illumina mouseRef-8 v1.0 expression beadchip 0005720484 - -
Illumina mouseRef-8 v1.0 expression beadchip 0005900168 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2594873 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2629897 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2594873 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2629897 - -
mouse-6 v1.1 (Illumina) scl074034.1_57-S - -
mouse-6 v1.1 (Illumina) scl0074034.1_260-S - -
Illumina Sentrix mouse-6.v1 bead array 5720484 - -
Illumina Sentrix mouse-6.v1 bead array 5900168 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl0074034.1_260-S - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl074034.1_57-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 5720484 - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 5900168 - -
MouseRef-8 v2 Expression BeadChip ILMN_2629897 - -
MouseRef-8 v2 Expression BeadChip ILMN_2594873 - -
MouseRef-8 v2 Expression BeadChip ILMN_2594872 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2594873 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2629897 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2594872 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0