This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03935509_s1
See other TMEM246 GE Assays ›
Gene Symbol:
Tmem246
Gene Name:
transmembrane protein 246
Gene Aliases:
2810432L12Rik, 9330170P15Rik, AI835809
Chromosome Location:
Chr.4: 49584506 - 49597870 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03935509_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tmem246
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK038607.1 - 1 748 103
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BI180689.1 - 1 346 Chr. 4 - 49596730 - 49597366 [-] on Build GRCm38 103

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream aldolase B,
fructose-bisphosphate
Aldob Aldo-2,
Aldo2,
BC016435
-47.2kb Sense Chr. 4 - 49535995 - 49549483 on NCBI Build GRCm38
Downstream ring finger protein 20 Rnf20 4833430L21Rik,
AW540162,
C79397,
mKIAA4116
34.2kb Anti-sense Chr. 4 - 49632011 - 49656886 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0