This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03935391_s1
See other KCNN4 GE Assays ›
Gene Symbol:
Kcnn4
Gene Name:
potassium intermediate/small conductance calcium-activated channel, subfamily N, member 4
Gene Aliases:
IK1, IKCA1, KCA4, KCa3.1, SK4, SKCas, mIKCa1
Chromosome Location:
Chr.7: 24369833 - 24385212 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03935391_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Kcnn4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK038253.1 - 3544 79
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BB166842.2 - 1 544 Chr. 7 - 24383054 - 24383686 [-] on Build GRCm38 79

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream Ly6/Plaur domain containing 5 Lypd5 2210003I03Rik -29.2kb Anti-sense Chr. 7 - 24349224 - 24353833 on NCBI Build GRCm38
Downstream smg-9 homolog,
nonsense mediated mRNA decay factor (C. elegans)
Smg9 1500002O20Rik,
AI429693,
N28092
15.2kb Anti-sense Chr. 7 - 24398910 - 24422777 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

MicroArray Probes:

ABI

Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 565191 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0