This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03934344_s1
See other 4632404H12RIK GE Assays ›
Gene Symbol:
4632404H12Rik
Gene Name:
RIKEN cDNA 4632404H12 gene
Gene Aliases:
-
Chromosome Location:
Chr.3: 89768430 - 89773233 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03934344_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 4632404H12Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK035453.1 - 1 352 79
BC063049.1 - 1 2928 79
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BB627495.1 - 1 352 Chr. 3 - 89769598 - 89770215 [-] on Build GRCm38 79

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream cholinergic receptor,
nicotinic,
beta polypeptide 2 (neuronal)
Chrnb2 Acrb-2,
Acrb2,
C030030P04Rik,
[b]2-nAchR
-3.8kb Sense Chr. 3 - 89753448 - 89764632 on NCBI Build GRCm38
Downstream ubiquitin-conjugating enzyme E2Q (putative) 1 Ube2q1 1110002C01Rik,
2310012M18Rik,
AI854014,
Gtap,
NICE-5,
PRO3094,
Ube2q
3.4kb Anti-sense Chr. 3 - 89773609 - 89783997 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

MicroArray Probes:

ABI

Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 757085 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header