This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03934214_s1
See other C530005A16RIK GE Assays ›
Gene Symbol:
C530005A16Rik
Gene Name:
RIKEN cDNA C530005A16 gene
Gene Aliases:
D930044c15rik
Chromosome Location:
Chr.4: 116589733 - 116597630 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03934214_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: C530005A16Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_029450.1 1 44 61
GenBank mRNA AK035052.1 - 1 49 61
AK039789.1 - 1 47 61
AK086654.1 - 1 45 61
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BB634126.1 - 1 42 Chr. 4 - 116597000 - 116597628 [-] on Build GRCm38 61
GenBankmRNA BB663430.1 - 1 42 Chr. 4 - 116596962 - 116597628 [-] on Build GRCm38 61
GenBankmRNA BB642790.1 - 1 42 Chr. 4 - 116597004 - 116597628 [-] on Build GRCm38 61
GenBankmRNA BY271235.1 - 1 Chr. 4 - 116597300 - 116597695 [+] on Build GRCm38 61
GenBankmRNA BY037189.1 - 1 35 Chr. 4 - 116592794 - 116597620 [-] on Build GRCm38 61
GenBankmRNA BB627295.1 - 1 42 Chr. 4 - 116597003 - 116597628 [-] on Build GRCm38 61
GenBankmRNA BY289133.1 - 1 44 Chr. 4 - 116597208 - 116597629 [-] on Build GRCm38 61

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream coiled-coil domain containing 17 Ccdc17 Anti-sense Chr. 4 - 116596730 - 116600266 on NCBI Build GRCm38
Downstream nuclear autoantigenic sperm protein (histone-binding) Nasp 5033430J04Rik,
AI131596,
AI317140,
D4Ertd767e,
Epcs32,
Nasp-T
3.4kb Sense Chr. 4 - 116601052 - 116627951 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400026703.mm.1 NR_029450.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400026703.mm.1 NR_029450.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400026703.mm.1 TC0400003342.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 718882 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0