This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03934199_s1
See other PPP1R18OS GE Assays ›
Gene Symbol:
Ppp1r18os
Gene Name:
protein phosphatase 1, regulatory subunit 18, opposite strand
Gene Aliases:
5530401N12Rik
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03934199_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ppp1r18os
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK035015.1 - 1 1851 86
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BB100689.2 - 1 596 Chr. 17 - 35859653 - 35860304 [-] on Build GRCm38 86

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream mediator of DNA damage checkpoint 1 Mdc1 6820401C03,
AA413496,
Nfbd1,
mKIAA0170
Anti-sense Chr. 17 - 35841490 - 35859670 on NCBI Build GRCm38
Downstream protein phosphatase 1,
regulatory subunit 18
Ppp1r18 2310014H01Rik,
AA407849,
AI450394
3.9kb Anti-sense Chr. 17 - 35865495 - 35875596 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

MicroArray Probes:

Agilent ABI

Array Name Probe IDs Targets Target Type
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2250154 AK035015.1 mouse_transcriptome
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 347365 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0