This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03933959_s1
See other DEP1 GE Assays ›
Gene Symbol:
Dep1
Gene Name:
diabetic embryopathy 1
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03933959_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dep1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK034614.1 - 1 1546 78

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream quaking Qk 1110003F05Rik,
1500005P18,
QkI,
l(17)-1Wis,
l17Wis1
Sense Chr. 17 - 10202856 - 10319361 on NCBI Build GRCm38
Downstream colon adenocarcinoma hypermethylated RNA Cahm B930003M22Rik 4.6kb Anti-sense Chr. 17 - 10319919 - 10321141 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2271058 AF032130.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2271058 AK034614.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P212334 AK034614.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P123439 - -
Array Name Probe IDs Targets Target Type
Affymetrix HT MG-430 PM Array Plate 1427602_PM_at - -
Affymetrix HT MG-430 PM Array Plate 1438892_PM_at - -
Mouse Genome 430 2.0 Array 1427602_PM_at - -
Mouse Genome 430 2.0 Array 1438892_at - -
Mouse Genome 430 2.0 Array 1427602_at - -
Mouse Genome 430 2.0 Array 1438892_PM_at - -
Mouse Expression Set 430 1427602_at - -
Mouse Expression Set 430 1427602_PM_at - -
Mouse Expression Set 430 1438892_at - -
Mouse Expression Set 430 1438892_PM_at - -
Mouse Genome 430A 2.0 Array 1427602_PM_at - -
Mouse Genome 430A 2.0 Array 1427602_at - -
Affymetrix HT Mouse Genome 430A Array 1427602_PM_at - -
Affymetrix HT Mouse Genome 430A Array 1427602_at - -
Affymetrix HT Mouse Genome 430B Array 1438892_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1438892_at - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0