This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03933360_s1
See other 4432414F05RIK GE Assays ›
Gene Symbol:
4432414F05Rik
Gene Name:
RIKEN cDNA 4432414F05 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03933360_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 4432414F05Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK014495.1 - 1 1145 68
AK033334.1 - 1 847 68

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream zinc finger protein 346 Zfp346 Jaz,
Znf346
Sense Chr. 13 - 55105271 - 55135071 on NCBI Build GRCm38
Downstream fibroblast growth factor receptor 4 Fgfr4 Fgfr-4 40.5kb Sense Chr. 13 - 55152818 - 55168759 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Affymetrix HT MG-430 PM Array Plate 1453976_PM_at - -
Mouse Genome 430 2.0 Array 1453976_at - -
Mouse Genome 430 2.0 Array 1453976_PM_at - -
Mouse Expression Set 430 1453976_PM_at - -
Mouse Expression Set 430 1453976_at - -
Affymetrix HT Mouse Genome 430B Array 1453976_at - -
Affymetrix HT Mouse Genome 430B Array 1453976_PM_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 872227 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 106420114 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0