This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03933206_m1
See other A730085K08RIK GE Assays ›
Gene Symbol:
A730085K08Rik
Gene Name:
RIKEN cDNA A730085K08 gene
Gene Aliases:
-
Chromosome Location:
Chr.9: 122382022 - 122398801 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03933206_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: A730085K08Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_045967.1 1 - 2 148 98
GenBank mRNA AK033049.1 - 1 - 2 146 98
AK043328.1 - 149 98
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BB624012.1 - 1 - 2 146 Chr. 9 - 122382064 - 122398798 [-] on Build GRCm38 98
GenBankmRNA AV360585.2 - 1 - 2 158 Chr. 9 - 122382066 - 122398798 [-] on Build GRCm38 98

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream abhydrolase domain containing 5 Abhd5 1300003D03Rik,
2010002J10Rik,
CDS,
CGI-58,
IECN5,
NCIE2
-8.0kb Anti-sense Chr. 9 - 122351539 - 122381524 on NCBI Build GRCm38
Downstream RIKEN cDNA 9530059O14 gene 9530059O14Rik 173.7kb Anti-sense Chr. 9 - 122572501 - 122580647 on NCBI Build GRCm38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900003287.mm.1

Set Membership:

Amplicon length between 86 and 100 Mus musculus Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900027710.mm.1 NR_045967.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900027710.mm.1 NR_045967.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900027710.mm.1 TC0900003287.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900027711.mm.1 NR_045967.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900027711.mm.1 NR_045967.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900027711.mm.1 TC0900003287.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0