This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03932514_s1
See other 1700012D14RIK GE Assays ›
Gene Symbol:
1700012D14Rik
Gene Name:
RIKEN cDNA 1700012D14 gene
Gene Aliases:
-
Chromosome Location:
Chr.7: 111117673 - 111122675 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03932514_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 1700012D14Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK031468.1 - 1 1146 95
AK047116.1 - 1 1146 95
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AV321778.2 - 1 471 Chr. 7 - 111121322 - 111121995 [-] on Build GRCm38 95
GenBankmRNA CO039024.1 - 1 Chr. 7 - 111121306 - 111122032 [+] on Build GRCm38 95
GenBankmRNA AI835017.1 - 1 Chr. 7 - 111121307 - 111121677 [+] on Build GRCm38 95
GenBankmRNA BB742271.1 - 1 196 Chr. 7 - 111121323 - 111121744 [-] on Build GRCm38 95

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream eukaryotic translation initiation factor 4,
gamma 2
Eif4g2 AA589388,
DAP-5,
E130105L11Rik,
Nat1,
Natm1,
p97
-38.3kb Sense Chr. 7 - 111067985 - 111083030 on NCBI Build GRCm38
Downstream UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 18 Galnt18 2900011G21Rik,
BC024988,
Galntl4
349.0kb Sense Chr. 7 - 111471656 - 111780067 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0