This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03931515_s1
See other E130101A01RIK GE Assays ›
Gene Symbol:
E130101A01Rik
Gene Name:
RIKEN cDNA E130101A01 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03931515_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: E130101A01Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK021363.1 - 1 247 87
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BQ555165.1 - 1 471 Chr. 13 - 81726675 - 81727267 [+] on Build GRCm38 87
GenBankmRNA CA890910.1 - 1 Chr. 13 - 81727084 - 81727732 [-] on Build GRCm38 87

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream metallo-beta-lactamase domain containing 2 Mblac2 2900024O10Rik Sense Chr. 13 - 81711417 - 81753275 on NCBI Build GRCm38
Downstream uncharacterized LOC105245566 LOC105245566 55.0kb Anti-sense Chr. 13 - 81782594 - 81783769 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

MicroArray Probes:

Illumina

Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 102340524 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0