This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03931471_s1
See other D730047N16RIK GE Assays ›
Gene Symbol:
D730047N16Rik
Gene Name:
RIKEN cDNA D730047N16 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03931471_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: D730047N16Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK021346.1 - 1 547 99
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY730938.1 - 1 536 Chr. 6 - 17578078 - 17578713 [+] on Build GRCm38 99

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream met proto-oncogene Met AI838057,
HGF,
HGFR,
Par4,
c-Met
-4.1kb Sense Chr. 6 - 17463349 - 17573980 on NCBI Build GRCm38
Downstream capping protein (actin filament) muscle Z-line,
alpha 2
Capza2 1110053K06Rik,
AW208808,
Cappa2
58.3kb Sense Chr. 6 - 17637098 - 17666534 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

MicroArray Probes:

Agilent ABI

Array Name Probe IDs Targets Target Type
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 880870 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 157685 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05822976 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24268503 - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 693076 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0