This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03931377_s1
See other C430039J01RIK GE Assays ›
Gene Symbol:
C430039J01Rik
Gene Name:
RIKEN cDNA C430039J01 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03931377_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: C430039J01Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK021246.1 - 1 353 73
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY729179.1 - 1 353 Chr. 7 - 116410141 - 116410803 [-] on Build GRCm38 73

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream phosphatidylinositol 3-kinase,
C2 domain containing,
alpha polypeptide
Pik3c2a Cpk-m,
PI3KC2
Sense Chr. 7 - 116337265 - 116444557 on NCBI Build GRCm38
Downstream nucleobindin 2 Nucb2 AI607786,
Calnuc,
Nefa,
Nesfatin-1
93.6kb Anti-sense Chr. 7 - 116504353 - 116540588 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

MicroArray Probes:

Agilent

Array Name Probe IDs Targets Target Type
Whole Mouse Genome G4122A A_52_P891100 AK021246.1 mouse_transcriptome

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0