This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03931355_s1
See other C430004N12RIK GE Assays ›
Gene Symbol:
C430004N12Rik
Gene Name:
RIKEN cDNA C430004N12 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03931355_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: C430004N12Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK021233.1 - 1 247 79
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY728938.1 - 1 244 Chr. X - 152750974 - 152751588 [-] on Build GRCm38 79

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream claudin 34A Cldn34a Gm7157 -186.7kb Anti-sense Chr. X - 152537528 - 152564210 on NCBI Build GRCm38
Downstream G protein-coupled receptor 143 Gpr143 Oa1 30.3kb Anti-sense Chr. X - 152781921 - 152808651 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

MicroArray Probes:

ABI Illumina

Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 621478 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 106980546 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header