This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03931344_s1
See other C330024E10RIK GE Assays ›
Gene Symbol:
C330024E10Rik
Gene Name:
RIKEN cDNA C330024E10 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03931344_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: C330024E10Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK021222.1 - 1 651 68

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream serine/threonine kinase 32A Stk32a A930015B13Rik,
YANK1
-42.5kb Anti-sense Chr. 18 - 43207697 - 43317481 on NCBI Build GRCm38
Downstream eukaryotic translation initiation factor 3,
subunit J2
Eif3j2 ENSMUSG00000043424,
Eif3j-2,
Gm9781,
eIF3 p35,
eIF3j-B
114.6kb Sense Chr. 18 - 43475418 - 43477836 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

MicroArray Probes:

Illumina

Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 100450131 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header